Torch Module Support Stand, PerkinElmer
10136-312
- Description:Torch module support stand
- Cat. No.:10136-312
- Supplier no.:N0770609
The free-standing support stand is used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV, and 8x00 ICP-OES series instruments.
A free-standing support stand to hold spare quick-change torch module or as a means of conveniently holding module when working inside the sample compartment.