Order Entry
United States
ContactUsLinkComponent
Sample Loops for Oil Samples, PerkinElmer
Sample Loops for Oil Samples, PerkinElmer
Catalog # 10139-522
Supplier:  PerkinElmer
undefined
Sample Loops for Oil Samples, PerkinElmer
Catalog # 10139-522
Supplier:  PerkinElmer
Supplier Number:  N0777594

Specifications

  • Ø int.:
    1.0 mm
  • Capacity:
    1.02 ml
  • Description:
    Sample loop for oil samples
  • Cat. No.:
    10139-522
  • Supplier no.:
    N0777594

Specifications

About this item

The PFA sample loop is used with the AXPRESS® PLUS valve/pump module for oils applications.

The assembly includes 2 flangeless connectors.