Specifications
- Description:Universal GemTip nebulizer
- Environmentally Preferable:
- Cat. No.:10138-020
- Supplier no.:N0370394
Specifications
About this item
This universal corrosion-resistant GemTip plastic body nebulizer is for use with AAnalyst® 100/200/300/400/700/800 and earlier AA series instruments. It is corrosion resistant and can be used to aspirate almost all solutions, even those solutions containing a high percentage of acid.
A complete GemTip plastic body nebulizer featuring viton O-rings and plastic needle with sapphire tip.
This assembly includes: capillary tubing, Qty.1 (97012-048), fine adjustment nut, Qty.1 (101680-080), gland nut, Qty.1 (97012-744), PTFE tubing, Qty.1 (97012-040), capillary assembly for universal GemTip nebulizers, Qty.1 (10138-026) O-Ring for capillary assembly, Qty.1 (101692-784), O-Ring for nebulizer body, Qty.1 (97012-136), body assembly for universal GemTip nebulizer, Qty.1 (10138-022), locking nut, Qty.1 (101675-032), spring, Qty.1, connector nut, Qty.1 (101675-030), magnet holder assembly, Qty.1 (101681-786).
The nebulizer is compatible with the PerkinElmer AAnalyst® 200/400*, AAnalyst®100/300/700/800, and Models 5100PC/5000/3300/3110/3100 AA instruments. Note: AAnalyst® 200/400 requires an end cap assembly (10138-018).