ContactUsLinkComponent
Phosphorus (P) Lumina™ Hollow Cathode Lamp, PerkinElmer
Phosphorus (P) Lumina™ Hollow Cathode Lamp, PerkinElmer
Catalog # 45603-324
Supplier:  PerkinElmer
undefined
Phosphorus (P) Lumina™ Hollow Cathode Lamp, PerkinElmer
Catalog # 45603-324
Supplier:  PerkinElmer
Supplier Number:  N3050155
Specifications
  • Description:
    Phosphorus (P) Lumina™ hollow cathode lamp
  • Cat. No.:
    45603-324
  • Supplier no.:
    N3050155
Specifications
About this item
Single Element Lumina™ hollow cathode lamp (HCL) for the detection of elemental Phosphorus (P). Series N30501XX Lumina™ 50 mm (2") diameter single-element lamps are designed to be used with our PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
Please note that this element requires the use of a nitrous oxide-acetylene flame.
Lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. Offers a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ hollow cathode lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime.