About this item
Eppendorf Rotors for Centrifuge 5420, precision for diverse applications. Choose from various capacities and speeds to tailor your centrifuge setup. Accessorize with adaptors and buckets, ensuring optimal sample processing in molecular biology and beyond.
- Precision-engineered for optimal sample separation
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
- Accessories (adaptors and buckets) further customize your centrifuge setup
- Aerosol-tight options available
Elevate your centrifugation capabilities with Eppendorf Rotors designed for Centrifuge 5420. Tailor your experiments with a range of capacities and speeds, offering flexibility for microcentrifugation or larger sample volumes. Accessorize your setup with adaptors and buckets, customizing for specific applications in molecular biology and beyond. Achieve precise sample separation, ensuring reproducible results in diverse workflows. Explore the comprehensive range to optimize your Centrifuge 5420 for reliable performance.
Specifications
- Catalog No:
- 76404-390
- 76404-392
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Max. RPM:
- 15060 rpm
- 15060 rpm
- Environmental Certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001