ContactUsLinkComponent
Leica HistoCore NANOCUT R Automated Research and Special Applications Rotary Microtome
Leica HistoCore NANOCUT R Automated Research and Special Applications Rotary Microtome
Catalog # 76404-250
undefined
Leica HistoCore NANOCUT R Automated Research and Special Applications Rotary Microtome
Catalog # 76404-250
Supplier Number:  14052458261
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    HistoCore NANOCUT R automated research and special applications rotary microtome
  • Cat. No.:
    76404-250
Specifications
About this item
When microtomy involves diverse samples in order to discover new breakthroughs in research projects, then NANOCUT R is the optimal solution. From biomedical research to industrial applications, NANOCUT R can cut virtually anything can be put in front of it.
  • Cut sections that can be prepared for analysis in light microscopy and electron microscopy - down to 250 nm (0.25 µm, semi-thin)
  • Operating humidity range of 20 to 80% maximum (non-condensing)
  • Maximum heat emission of 340 J/s
Preserve sample integrity when performing semi-thin sectioning with the special spindle and expanded slow speed adjustment. Precisely stop the sample in the object head's optimal position for photo-capture of the cut sample surface when running 3-dimensional reconstruction programs.
Operating temperature: 18...35 °C