ContactUsLinkComponent
Tweezers, Component Handling, Excelta Corp®
Tweezers, Component Handling, Excelta Corp®
Catalog # 89411-708
Supplier:  Excelta
undefined
Tweezers, Component Handling, Excelta Corp®
Catalog # 89411-708
Supplier:  Excelta
Supplier Number:  AH-SA-SE
Specifications
  • Description:
    Tweezers, arrow head, 1 star quality
  • Material:
    Stainless steel, anti-magnetic
  • Curvature:
    Straight
  • Cat. No.:
    89411-708
  • Overall length:
    125 mm (5")
  • Supplier no.:
    AH-SA-SE
Specifications
About this item
Unique triangle shape for handling large items.
  • Tip designed for handle large items
  • Finger serrations for 'non-slip' use
This straight strong serrated tipped 4.25" tweezer is made of AISI 303/304 stainless steel to protect from oxidation. It features finger serrations for 'non-slip' control. It's unique triangle shape makes it very strong.