Order Entry
United States
ContactUsLinkComponent
Biotin Conjugate Collagen Hybridizing Peptide (Biotin), Advanced Biomatrix
Biotin Conjugate Collagen Hybridizing Peptide (Biotin), Advanced Biomatrix
Catalog # 103792-904
CAS Number:  
undefined
Biotin Conjugate Collagen Hybridizing Peptide (Biotin), Advanced Biomatrix
Catalog # 103792-904
Supplier Number:  5265-60UG
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Biotin Conjugate Collagen Hybridizing Peptide (Biotin)
  • Storage Conditions:
    –20 °C as powder for long term storage. 4 °C after reconstitution in water. Protect from light.
  • Solubility:
    Aqueous Buffers
  • Cat. No.:
    103792-904

Specifications

About this item

CHP can specifically bind to denatured collagen.

  • Useful in histology, in vivo, and 3D cell culture
  • Source: Synthetic

The triple helix is the hallmark protein structure of collagen. During tissue remodeling, the triple helical collagen molecules are degraded by specific proteases (e.g., MMP or cathepsin K) and become unfolded at body temperature. The Collagen Hybridizing Peptide (CHP) is a synthetic peptide that can specifically bind to such denatured collagen strands through hydrogen bonding, both in histology, in vivo, and in vitro (3D cell culture). By sharing the structural motif and the Gly-X-Y repeating sequence of natural collagen, CHP has a strong capability to hybridize with denatured collagen strands, in a fashion that is similar to a DNA fragment annealing to its complimentary DNA strand during PCR. CHP is an extremely specific probe for unfolded collagen molecules: it has negligible affinity to intact collagen molecules due to the lack of binding sites; it is also inert towards non-specific binding because of its neutral and hydrophilic nature.