To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/Peptide Type:Native
- Source:Hide
- Species:Bovine
- Size:20 ml
- Endotoxin Content:<10.0 EU/mL
- Form:Solution
- Protein/Peptide Name:Collagen, Type I
- Cat. No.:103792-884
Specifications
About this item
TeloCol®-10 is 20 mL of 10 mg/ml, type I bovine telocollagen solution for 2D coatings or 3D hydrogels. TeloCol®-10 has a high viscosity and concentration, and can form firm hydrogels.
- Highly purified type I telocollagen
- High concentration
- Lot to Lot consistency
TeloCol®-10 bovine collagen solution is derived from an acid extraction process yielding a telopeptide-intact collagen. The pro-peptide regions at both ends of the collagen chain, N- and C-telopeptide regions, are maintained.
TeloCol®-10 collagen is approximately 95% Type I collagen with the remainder being comprised of Type III collagen. Each product includes a bottle containing 20 ml of collagen solution. This product is supplied at a concentration of approximately 10 mg/ml. TeloCol®-10 is sterile filtered and provided in user-friendly packaging for use and storage.
TeloCol®-10 is ideal for coating of surfaces, providing preparation of thin layers for culturing cells, or use as a solid gel.