ContactUsLinkComponent
Stoppers/Adapters, Extraction Nut, 3/4" NPT, Chemglass
Stoppers/Adapters, Extraction Nut, 3/4" NPT, Chemglass
Catalog # 76333-406
Supplier:  Chemglass
undefined
Stoppers/Adapters, Extraction Nut, 3/4" NPT, Chemglass
Catalog # 76333-406
Supplier:  Chemglass
Supplier Number:  CG-3002-K-48
Specifications
  • Description:
    45/50 ptfe stopper, 3/4" npt, complete with extraction nut
  • Cat. No.:
    76333-406
Specifications
About this item
PTFE stopper adapter is similar to CG-3002-L but with the addition of one female NPT port in the center.
The optional PTFE compression fittings provide a convenient way to insert probes or connect PFA tubing or FEP tubing to reactors or flasks. PTFE stopper has an extraction nut permitting easy removal of the stopper from the glass outer joint.