Order Entry
United States
ContactUsLinkComponent
Replacement Stirrer Shaft, 25.4 mm, for Reactor Size 30 L, Chemglass
Replacement Stirrer Shaft, 25.4 mm, for Reactor Size 30 L, Chemglass
Catalog # 76333-722
Supplier:  Chemglass
undefined
Replacement Stirrer Shaft, 25.4 mm, for Reactor Size 30 L, Chemglass
Catalog # 76333-722
Supplier:  Chemglass
Supplier Number:  CG-2097-33

Specifications

  • Description:
    Replacement 25.4 mm stirrer shaft, reactor size: 30 L, fits support frame: cg-1968-x-12, overall Length 40.9"
  • Cat. No.:
    76333-722

Specifications

About this item

Precision ground and polished stirrer shaft for use with Chemglass process reactors.

Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a ³/₈" wide flat.

Caution: Note: cg-2097-75 and cg-2097-100 are made from heavy wall tubing and have indents for agitator positioning. Indents are spaced every 10" from the bottom agitator.