ContactUsLinkComponent
Heavy Duty Cart, For use with Huber Unistat models 405 and 705 Circulators, Chemglass
Catalog # 76333-636
Supplier:  Chemglass
undefined
Heavy Duty Cart, For use with Huber Unistat models 405 and 705 Circulators, Chemglass
Catalog # 76333-636
Supplier:  Chemglass
Supplier Number:  CG-1976-H100
Specifications
  • Description:
    Cart, heavy duty, circulator, supplied with 8' tie down strap, for use with unistat 405 or 705
  • Cat. No.:
    76333-636
Specifications
About this item
Heavy duty cart for use with huber unistat models 405 and 705 circulators.
Cart raises the circulators so that the controllers are easily viewed and may make it possible to use shorter hoses. Cart will support up to 500 lbs., has a light gray powder coated finish with two locking casters and supplied with one 8' nylon tie down strap.