ContactUsLinkComponent
Digital Display Meters/Monitor, for use with CG-1872 pH Electrodes with an Integrated PT100 Sensor, Chemglass
  76333-628
 :  Chemglass
undefined
Digital Display Meters/Monitor, for use with CG-1872 pH Electrodes with an Integrated PT100 Sensor, Chemglass
  76333-628
 :  Chemglass
 :  CG-1872-M-40
  • Description:
    pH meter, stainless steel enclosure, digital display, 115v, with output to record data, supplied with cg-1872-m-25
  • Cat. No.:
    76333-628
Digital display meters/monitors for use with CG-1872 pH electrodes with an integrated PT100 sensor for temperature correction.
Connection is made using a standard BNC probe connector with a 3-pin banana connector plug for RTD. CG-1872-M-20 and CG-1872-M-40 meters are supplied with CG-1872-M-25 adapter cables.