ContactUsLinkComponent
Copper Torch Ignitor Tape for Optima™ 8x00, PerkinElmer
Copper Torch Ignitor Tape for Optima™ 8x00, PerkinElmer
  10130-472
undefined
Copper Torch Ignitor Tape for Optima™ 8x00, PerkinElmer
  10130-472
 :  N0781097
  • Description:
    Copper torch ignitor tape for optima™ 8x00
  • Cat. No.:
    10130-472
  • Packaging:
    1 Roll
  • Supplier no.:
    N0781097
This copper foil torch ignitor tape is used with Optima™ 8x00 ICP-OES series instruments and can be used for both Optima™ and HybridXLT™ torches.
  • Roll of tape includes 50 pieces
This copper ignitor tape is also included in the Scott/Cross-Flow II Sample Introduction Kit (N0780616) and the Cyclonic/Concentric Sample Introduction Kit (N0780617) for Optima™ 8x00.