Impact Bead for High Sensitivity Plastic Body Nebulizers, PerkinElmer
Catalog # 97012-848
Supplier: PerkinElmer
Specifications
- Description:Impact bead for high-sensitivity nebulizers
- Cat. No.:97012-848
- Supplier no.:B0505086
Specifications
About this item
This ceramic impact bead is for use with AAnalyst® 100/300/200/400/700/800 and earlier AA series instruments. It is designed for use in High Sensitivity Plastic Body Nebulizers.
This impact bead is for use with the Organics High Sensitivity Nebulizer Complete Assembly with Pt/Ir Capillary for AAnalyst® (97013-652) and the Organics High Sensitivity Nebulizer Complete Assembly with Tantalum Capillary for AAnalyst (10139-052).