Torch Adapter with Auxiliary Port, PerkinElmer
Catalog # 10139-148
Supplier: PerkinElmer
Specifications
- Description:Torch adapter with auxillary port
- Cat. No.:10139-148
- Supplier no.:N0774070
Specifications
About this item
This torch adapter, with an auxiliary port for oxygen addition, is used with PerkinElmer Optima™ 4300 V, 5300 V, and 7300 V ICP-OES series instruments.
This torch adapter with auxiliary oxygen port is also included in the V Oxygen addition kit for optima™ 4300 V/5300 V/7300 V (N0770965).