ContactUsLinkComponent
SeaSpray™ Quartz Nebulizer for NexION®, ELAN®, and Optima® Instruments, PerkinElmer
SeaSpray™ Quartz Nebulizer for NexION®, ELAN®, and Optima® Instruments, PerkinElmer
Catalog # 10139-146
Supplier:  PerkinElmer
undefined
SeaSpray™ Quartz Nebulizer for NexION®, ELAN®, and Optima® Instruments, PerkinElmer
Catalog # 10139-146
Supplier:  PerkinElmer
Supplier Number:  N0774069
Specifications
  • Description:
    SeaSpray™ quartz nebulizer for ICP-MS and ICP-OES
  • Cat. No.:
    10139-146
  • Supplier no.:
    N0774069
Specifications
About this item
This concentric SeaSpray™ Quartz Nebulizer is designed for use with PerkinElmer ELAN® 5000/6x00/9000/DRCs and NexION® 300/350/1000/2000 ICP-MS series of ICP-MS instruments. It offers freedom from clogging while nebulizing solutions near the limit of solubility of most mineral salts and conferring significant sensitivity gains.
  • 1.0 L/min. Argon flow rate
  • 1 mL/min. uptake rate
  • Ideal for samples with high dissolved solids
  • Outstanding nebulization efficiency for trace level analysis
All SeaSpray™ Nebulizers are supplied with the EzyLok Argon connector.