Vernier Go Direct® Physics Starter Package
470319-994
- Description:Go Direct® Physics starter package
- Cat. No.:470319-994
This includes Go Direct® Motion detector, Go Direct® Force and acceleration sensor, Go Direct® Voltage probe, Go Direct® Current probe.
Software: Graphical Analysis™ 4 for Chrome™, iOS, Android™, Windows®, and macOS®. Collect, share, and analyze sensor data with our free software. Graphical analysis four app facilitates student understanding with real-time graphs of experimental data and intuitive analysis tools.