ContactUsLinkComponent
Vernier Go Direct® Physics Standard Package
Vernier Go Direct® Physics Standard Package
Catalog # 470319-992
undefined
Vernier Go Direct® Physics Standard Package
Catalog # 470319-992
Supplier Number:  GDP-PHY-DX
Specifications
  • Description:
    Go Direct® Physics standard package
  • Cat. No.:
    470319-992
Specifications
About this item
This includes Go Direct® Motion detector, Go Direct® Force and acceleration sensor, Go Direct® Voltage probe, Go Direct® Photogate, Go Direct® Current probe (2), ultra pulley attachment, picket fence, Go Direct® Acceleration sensor, Go Direct® Sound sensor, Go Direct® Light and color sensor, Go Direct® 3-Axis magnetic field sensor.
Software: Graphical Analysis™ 4 for Chrome™, iOS, Android™, Windows®, and macOS®. Collect, share, and analyze sensor data with our free software. Graphical analysis four app facilitates student understanding with real-time graphs of experimental data and intuitive analysis tools.