ContactUsLinkComponent
Accessories for 4000 Series Control Panels, Fixed Systems Gas Detection
Accessories for 4000 Series Control Panels, Fixed Systems Gas Detection
Catalog # 76334-820
undefined
Accessories for 4000 Series Control Panels, Fixed Systems Gas Detection
Catalog # 76334-820
Supplier Number:  2603-020
Specifications
  • Description:
    Regulator - Fixed Flow, 0.5 LPM, Gauge-yes, on/off-yes, 34L Steel, Exotic Gases-no (not for use with Exotic Gases)[for use with 34 L steel disposable cylinders with female (outside) thread]
  • Cat. No.:
    76334-820