ContactUsLinkComponent
Avantor® ACE®, Preparative Column Coupler
Avantor® ACE®, Preparative Column Coupler
Catalog # 76392-066
Supplier:  Avantor
undefined
Avantor® ACE®, Preparative Column Coupler
Catalog # 76392-066
Supplier:  Avantor
Supplier Number:  C0002
Specifications
  • Description:
    Column coupler (for use with H0009)
  • Cat. No.:
    76392-066
Specifications
About this item
Column coupler made from stainless steel/PEEK suitable for connecting preparative columns to stand-alone guard cartridge holder, or for making similar HPLC connections.
Ideally suited to connect 30 mm id preparative columns to H0009 cartridge holder.