Avantor® Partisphere C18, HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
Avantor® Partisphere® was one of the first commercially available spherical silicas and continues to provide reproducible, high efficiency separations.
- Spherical 5 µm porous silica
- Unique void sealing cartridge hardware
- Convenient and easy to use - hand tightened fittings
- Increased column lifetime
Stainless steel column hardware.
Specifications
- Catalog No:
- 76398-276
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental Certification:
- ISO 14001:2015 (530840 UM15)