VeriSpec® ICP Multi-Element Standard 23, RICCA Chemical Company
Catalog # RCRV010716100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Matrix:5% HNO₃/tr HF
- Pack type:Natural LDPE bottle
- Description:VeriSpec® ICP multi-element standard 23
- Size:100 ml
- Cat. No.:RCRV010716100N
- Supplier no.:RV010716-100N
Specifications
About this item
23 component standard.
15K ppm: Mg; 8K ppm: Na;3.5K ppm: Ca, 3000 ppm: K; 100 ppb: B, Fe, Mo, Sr; 50 ppb: As, Ba, Ni, V, Zn; 30 ppm: Mn; 25 ppm: Co, Pb; 20 ppm: Be, Cd, Cr, Cu; 10 ppm: Bi, Se, Tl.
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility