Order Entry
United States
ContactUsLinkComponent
VeriSpec® ICP Multi-Element Standard 9, RICCA Chemical Company
VeriSpec® ICP Multi-Element Standard 9, RICCA Chemical Company
Catalog # RCRV010715100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® ICP Multi-Element Standard 9, RICCA Chemical Company
Catalog # RCRV010715100N
Supplier:  Ricca Chemical
Supplier Number:  RV010715-100N
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Natural LDPE bottle
  • Description:
    VeriSpec® ICP multi-element 9
  • Size:
    100 ml
  • Cat. No.:
    RCRV010715100N
  • Supplier no.:
    RV010715-100N

Specifications

About this item

9 component standard.

100 ppm: As, Be, Pb, Cd, Cr, Ni, Hg, Se, Tl

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility