Avantor® Hichrom, Column End Plug
Catalog # 76399-014
Supplier: Avantor
Specifications
- Description:Column end plug
- Cat. No.:76399-014
Specifications
About this item
These polymer column end plugs are used to prevent solvent evaporation when the columns are not in use.
- Fingertight design
- Extend column lifetime
- Fits any size column
- Seal column for storage