ContactUsLinkComponent
VeriSpec® ICP Multi-Element Standard 7, RICCA Chemical Company
VeriSpec® ICP Multi-Element Standard 7, RICCA Chemical Company
  RCRV010702100N
 :  
undefined
VeriSpec® ICP Multi-Element Standard 7, RICCA Chemical Company
  RCRV010702100N
 :  RV010702-100N
 :  
  • Matrix:
    5% HNO₃/tr HF
  • Pack type:
    Natural LDPE bottle
  • Description:
    VeriSpec® ICP multi-element standard 7
  • Size:
    100 ml
  • Cat. No.:
    RCRV010702100N
  • Supplier no.:
    RV010702-100N
7 component standard.
1000 ppm: K
100 ppm: Ag, Al, B, Ba, Na
50 ppm: Si
 : Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility