VeriSpec® ICP Multi-Element Standard 17, RICCA Chemical Company
Catalog # RCRV010682100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Matrix:5% HNO₃
- Pack type:Natural LDPE bottle
- Description:VeriSpec® ICP multi-element standard 17
- Size:100 ml
- Cat. No.:RCRV010682100N
- Supplier no.:RV010682-100N
Specifications
About this item
17 component standard.
10 ppm: Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility