Order Entry
United States
ContactUsLinkComponent
VeriSpec® ICP Multi-Element Standard 18, RICCA Chemical Company
VeriSpec® ICP Multi-Element Standard 18, RICCA Chemical Company
Catalog # RCRV010676100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® ICP Multi-Element Standard 18, RICCA Chemical Company
Catalog # RCRV010676100N
Supplier:  Ricca Chemical
Supplier Number:  RV010676-100N
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Natural poly bottle
  • Description:
    VeriSpec® ICP multi-element standard 18
  • Size:
    100 ml
  • Cat. No.:
    RCRV010676100N
  • Supplier no.:
    RV010676-100N

Specifications

About this item

18 component standard.

100 ppm: Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Ni, Pb, Se, Th, Tl, U, V, Zn

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility