ContactUsLinkComponent
Avantor® Hichrom, Column Coupler, PEEK
  76399-124
 :  Avantor
undefined
Avantor® Hichrom, Column Coupler, PEEK
  76399-124
 :  Avantor
 :  HI-1112
  • Description:
    Male-male column coupler, 12.7 mm
  • Material:
    PEEK
  • Cat. No.:
    76399-124
Ideal for connecting analytical columns.