Order Entry
United States
ContactUsLinkComponent
Optic USB Base Station
Optic USB Base Station
Catalog # 470323-564
undefined
Optic USB Base Station
Catalog # 470323-564
Supplier Number:  BASE-U-4

Specifications

  • Interface:
    USB
  • Description:
    HOBO optic USB base station, operating temperature range 0...50 °C (32...122 °F)
  • Cat. No.:
    470323-564

Specifications

About this item

The Universal Optic USB Base Station and couplers are used to offload data from any HOBO® data logger with an Optic USB interface.

  • Offload data from any HOBO® data logger with an Optic USB interface
  • Couplers are included
  • Cable length 183 cm (6')
  • Offloads full 64K logger in 30 seconds
Delivery: Couplers for all compatible data loggers are included with the BASE-U-4. Couplers are included for compatibility with: UA Pendants, U20 water level data loggers, the U22 Water Temp Pro v2, our UTBI TidbiT, U24 Conductivit,y and U23 data loggers.