VeriSpec® Multi-Element Standard 4, RICCA Chemical Company
Catalog # RCRV010899100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:1000 ppm
- Matrix:5% HNO₃
- Description:VeriSpec® multi-element standard 4
- Size:100 ml
- Cat. No.:RCRV010899100N
- Supplier no.:RV010899-100N
Specifications
About this item
Four components standard.
1000 ppm: Ca, K, Mg, Na
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility