VeriSpec® Multi-Element Standard 26, RICCA Chemical Company
Catalog # RCRV010836100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:100 ppm
- Matrix:5% HNO₃/tr. HF/tr. Tartaric acid
- Description:VeriSpec® multi-element standard 26
- Size:100 ml
- Cat. No.:RCRV010836100N
- Supplier no.:RV010836-100N
Specifications
About this item
26 components standard.
100 ppm: Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn.
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility