Order Entry
United States
ContactUsLinkComponent
Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II) photoinitiator, for visible light crosslinking
Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II) photoinitiator, for visible light crosslinking
Catalog # 103792-886
Supplier:  Advanced Biomatrix
CAS Number:  
undefined
Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II) photoinitiator, for visible light crosslinking
Catalog # 103792-886
Supplier:  Advanced Biomatrix
Supplier Number:  5248-1KIT
CAS Number:  

Specifications

  • Size:
    1 kit
  • Cat. No.:
    103792-886

Specifications

About this item

 :
 :796.334 g/mol
Storage Temperature:Ambient
CAS Number:92361-49-4
Formula:C₄₆H₄₅ClP₂Ru
MDL Number:MFCD09038426