Order Entry
United States
ContactUsLinkComponent
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) photoinitiator, not sterile 405 nm
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) photoinitiator, not sterile 405 nm
Catalog # 103792-908
CAS Number:  
undefined
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) photoinitiator, not sterile 405 nm
Catalog # 103792-908
Supplier Number:  5269-500MG
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Size:
    500 mg
  • Cat. No.:
    103792-908

Specifications

About this item

 :
 :294.21 g/mol
Storage Temperature:Refrigerator
CAS Number:85073-19-4
Formula:C₁₆H₁₆LiO₃P
MDL Number:MFCD29472537