ContactUsLinkComponent
Dual Combination Wrist Strap and Footwear Tester, SCS, Desco Industries
Dual Combination Wrist Strap and Footwear Tester, SCS, Desco Industries
Catalog # 76382-938
Supplier:  Desco
undefined
Dual Combination Wrist Strap and Footwear Tester, SCS, Desco Industries
Catalog # 76382-938
Supplier:  Desco
Supplier Number:  770758
Specifications
  • Description:
    Dual combination tester
  • Cat. No.:
    76382-938
Specifications
About this item
This Dual Combination Wrist Strap and Footwear Tester designed to test the operator’s path-to-ground, and to determine that the operator’s personnel grounding devices (wrist straps, foot grounders, ESD shoes, smocks) are functioning correctly.
  • Compatible with single-wire and dual-wire wrist straps
  • Simultaneous testing of wrist strap and footwear in less than three seconds
  • Solid-state test switch with no moving parts
  • Split foot plate for testing foot grounders (or ESD shoes)
  • Powered by 30 V test signal
  • Relay for use with automatic doors or other locking systems
  • Use the SCS 770759 stand as an alternate installation method for the dual combination tester
  • 100/240 VAC power adapter with interchangeable plugs
This tester verifies the functionality of personnel grounding devices (wrist straps, foot grounders, ESD shoes, smocks). Three independent tests are performed simultaneously on the wrist strap and each foot. Green lights indicate a pass condition. Red and yellow lights plus an audible alarm indicate when the wrist strap and/or footwear (left or right) have failed. DIP switches may be accessed for test limit preferences.
Access control to an ESD protected area can be further enforced by using the relay terminal on the dual combination tester and connecting it to an electronic door lock or turnstile to grant access only to those who have passed their personal grounding device tests.
Wrist strap: 750 kilohms and 10 megohms
Footwear: 750 kilohms and 35 megohms
Certifications: Calibrated to NIST standards and includes a certificate.
Ordering information: Accessories not included. Optional stand sold separately.