ContactUsLinkComponent
Autosampler Probe with Filtered Tip, PerkinElmer
Autosampler Probe with Filtered Tip, PerkinElmer
Catalog # 10138-118
Supplier:  PerkinElmer
undefined
Autosampler Probe with Filtered Tip, PerkinElmer
Catalog # 10138-118
Supplier:  PerkinElmer
Supplier Number:  N0771529
Specifications
  • Description:
    Stainless steel sample probe with filtered tip
  • Cat. No.:
    10138-118
  • Supplier no.:
    N0771529
Specifications
About this item
Stainless steel sample probe for autosamplers ASX-260, ASX-280, ASX-520, ASX-520HS, ASX-560, EXR-8, XLR-8, XLR-860, ASX-1400 and ASX-1600.
The stainless steel sample probe features a screen tip and is used in oils applications. The probe includes the capillary tube fitting, although does not include capillary tubing.