ContactUsLinkComponent
Torch Mounting Block Assembly for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Torch Mounting Block Assembly for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Catalog # 10136-308
Supplier:  PerkinElmer
undefined
Torch Mounting Block Assembly for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Catalog # 10136-308
Supplier:  PerkinElmer
Supplier Number:  N0770600
Specifications
  • Description:
    Torch module mounting block for Optima
  • Cat. No.:
    10136-308
  • Supplier no.:
    N0770600
Specifications
About this item
Torch mounting block assembly for PerkinElmer Optima ICP-OES instruments.
  • Quick-release mechanism permits fast changing of sample introduction components
  • Compatible with a variety of torches and sample introduction systems for Optima ICP-OES instruments
The torch module mounting block is a sample introduction component used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV and 8x00 ICP-OES series instruments.
It is used in the Quick-Change Torch Module with Cyclonic Spray Chamber and Concentric Nebulizer for Optima 2x00/4x00/5x00/7x00 (97013-832) and for Optima 8x00, and the Quick-Change Torch Module with Scott Spray Chamber and GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 (10136-310).
Ordering information: Torch, injector, spray chamber or nebulizer must be purchased separately.