Order Entry
United States
ContactUsLinkComponent
SeaSpray™ Glass Nebulizer, 2.0 ml/min, for PerkinElmer Avio and Optima ICP-OES Instruments, PerkinElmer
SeaSpray™ Glass Nebulizer, 2.0 ml/min, for PerkinElmer Avio and Optima ICP-OES Instruments, PerkinElmer
Catalog # 10138-174
Supplier:  PerkinElmer
undefined
SeaSpray™ Glass Nebulizer, 2.0 ml/min, for PerkinElmer Avio and Optima ICP-OES Instruments, PerkinElmer
Catalog # 10138-174
Supplier:  PerkinElmer
Supplier Number:  N0811305

Specifications

  • Description:
    SeaSpray™ glass nebulizer
  • Material:
    Glass
  • Cat. No.:
    10138-174
  • Supplier no.:
    N0775345

Specifications

About this item

The SeaSpray™ concentric glass nebulizer has a 2 ml/min uptake rate, a 0.7 L/min air flow rate.

  • Ideal for samples with high dissolved solids
  • Outstanding nebulization efficiency for trace-level analyses
  • Freedom from clogging while nebulizing solutions to the limit of solubility of most mineral salts and conferring significant sensitivity gains

Includes UniFit Connector with 0.75×1.3×700 mm (I.D×O.D×L) long tubing (101675-616) and EzyLok Connector Kit (10139-382).

The nebulizer is compatible with PerkinElmer Optima 4x00 DV, 5x00 DV, 7x00 DV, 8x00 and Avio 200 and 500 ICP-OES instruments.