Three-Shelf Collapsible Wire Utility Cart, Luxor
76203-856
: Luxor
- Description:Utility cart
- Overall Dimensions:Overall: 33.75W×19.5D×39.5"H;
Clearance between shelves: 9.875" - Cat. No.:76203-856
Experience the durability and mobility of the Luxor Three-Shelf Collapsible Wire Utility Cart, a uniquely flexible and versatile unit that's perfect for offices, warehouses, restaurants, kitchens and hospitality environments.
- Shelves easily collapse for storage or transportation
- Constructed of steel with a polished chrome finish
- Single-grab handle for safe transport
- Rolling cart with full-swivel 4" Thermoplastic Rubber (TPR) casters, two with locking brakes
- Minimal assembly required – simply attach casters
- Holds up to 300 lbs. (100 lbs. per shelf), evenly distributed
Its open wire design with an anti-corrosion chrome finish increases ventilation and reduces dust, while its three large shelves provide plenty of room for storing a variety of items. It easily folds down to a space-saving footprint for storage or transport. This steel utility cart is shipped in one box and only requires attaching casters and locking the three shelves in place for assembly.