- Description:Stand up desk
- Cat. No.:76203-782
Quickly deliver collaborative and productive comfort to your workday by seamlessly alternating between sitting and standing with the Luxor pneumatic adjustable height standing desk.
- Electric push button control adjusts desk from 26 to 51.6"
- Generous 47.25×29.5" work surface provides plenty of room for multiple monitors
- Sturdy steel frame with 256 lbs. lift capacity
- Three-stage, telescoping leg system lifts heavier loads and allows lower and higher height adjustments
- Programmable memory keypad with up/down arrows store two preferred heights
- Anti-collision mechanism prevents lowering the desk on top of objects in its path
- Seven years warranty on the motors
Luxor electric height adjustable standing desk delivers exceptional performance at the push of a button. The two motors quietly power the three-stage leg columns to quickly raise and lower the desk at an impressive rate of 1.2" per second while a programmable memory keypad will store two customized settings for preferred user heights.
The solid steel frame features a telescoping leg system that offers greater adjustability while the spacious 48" desktop accommodates multiple monitors.
Whether used in a business or home office setting, this electric stand up desk delivers the health benefits of standing while increasing productivity throughout the workday.
Overall: W47.25×D29.5D×H26 - 51.6";
Desktop: W47.25×D29.5";
Cord: 9'