Drum Plug Socket for Round Head, ³/₄", Spark Resistant, Labelmaster
103258-076
- Material:Bronze
- Cat. No.:103258-076
- Weight:0.5 lbs.
- Supplier no.:KSOK75R
Get the proper fit using ¾" Drum Plug Sockets.
- Bronze, spark-resistant sockets
- May be used on round head plugs
- Fits ⅜" drives on a manual ratchet or manual torque wrench
Be sure to follow the closing instructions for sealing containers in order to be in compliance with UN torque requirements.