MICROCHEM® AlphaTec® Overboots for AlphaTec® 4000 Coveralls, Ansell
Catalog # 76214-708
Supplier: Ansell Healthcare
Specifications
- Shoe size:8–12
- Color:Green
- Cat. No.:76214-708
Specifications
About this item
Multi-layer barrier laminate construction.
- Welded seams
- Elastic to top of boot
- Tie fastening
Model 406 overboots are designed for use with the MICROCHEM® by AlphaTec® 4000 coveralls.
Packaging: Packaged 150 pairs per case.