Order Entry
United States
ContactUsLinkComponent
Air Marking Label 'Net/Gross Quantity', Labelmaster
Air Marking Label 'Net/Gross Quantity', Labelmaster
Catalog # 101377-574
Supplier:  Label Master
Air Marking Label 'Net/Gross Quantity', Labelmaster
Catalog # 101377-574
Supplier:  Label Master
Supplier Number:  L120

Specifications

  • Description:
    Air marking label
  • Dimensions:
    25.4×101.6 mm (1×4")
  • Material:
    Semi-gloss paper
  • Thickness:
    5.85 mil.
  • Cat. No.:
    101377-574
  • Legend:
    Net/Gross Quantity
  • Supplier no.:
    L120

Specifications

About this item

Inform shipping personnel of package details with Net/Gross Quantity Air Markings.

  • Ample space provided to fill in information
  • Made of paper
  • Outstanding chemical and abrasion resistance
  • Strong adhesion to corrugated boxes
  • Helps maintain regulatory compliance

These labels help satisfy the international air transport regulations requirement to show net quantity or gross weight on a package.