Order Entry
United States
ContactUsLinkComponent
Labels 'Dangerous When Wet', Labelmaster
Labels 'Dangerous When Wet', Labelmaster
Catalog # 101376-372
Supplier:  Label Master
Labels 'Dangerous When Wet', Labelmaster
Catalog # 101376-372
Supplier:  Label Master
Supplier Number:  HML23

Specifications

  • Description:
    Preprinted labels
  • Dimensions:
    100×100 mm (3.94×3.94")
  • Material:
    Semi-gloss paper
  • Thickness:
    5.85 mil.
  • Cat. No.:
    101376-372
  • Packaging:
    On roll
  • Legend:
    Dangerous When Wet
  • Supplier no.:
    HML23

Specifications

About this item

Employ a fool-proof shipping label solution with Worded Dangerous When Wet labels, revised to meet the design requirements of 49 CFR 172.407.

  • Printed with light-fast inks for fade resistance
  • Adhesives are formulated for use in various climates and environments
  • Once affixed, labels won't fade or fall off
  • Outstanding chemical and abrasion resistance

In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.