Order Entry
United States
Orders LinkContactUsLinkComponent
Special Waste Labels, Labelmaster

Special Waste Labels, Labelmaster

Special waste labels, PVC-free film, Special Waste, 152.4×152.4 mm (6×6"), 7.1 mil.
Catalog # 101376-798
Marketsource
Supplier:   Label Master
Supplier #:  PWMV
Special Waste Labels, Labelmaster
Catalog # 101376-798
Supplier:  Label Master
Marketsource
Special Waste Labels, Labelmaster

Special waste labels, PVC-free film, Special Waste, 152.4×152.4 mm (6×6"), 7.1 mil.

Price
$64.89
Log in for availability
Pack of 100
Minimum order quantity is 1
Product Details & Documents

Avoid potential regulatory violations when shipping solid waste by using Special Waste labels.

  • Constructed of rugged, flexible PVC-free vinyl
  • Automotive-grade ink
  • Specially formulated adhesives for use in various climates and environments

Ruled lines allow users to fill in the information necessary for marking a container for transport in compliance with the DOT and EPA regulations. These include all the information necessary for marking a container of special waste.

Specifications
  • Description:
    Special waste labels
  • Dimensions:
    152.4×152.4 mm (6×6")
  • Material:
    PVC-free film
  • Thickness:
    7.1 mil.
  • Legend:
    Special Waste