Order Entry
United States
ContactUsLinkComponent
Labels 'Protect From Freezing', Labelmaster
Labels 'Protect From Freezing', Labelmaster
Catalog # 101376-632
Supplier:  Label Master
Labels 'Protect From Freezing', Labelmaster
Catalog # 101376-632
Supplier:  Label Master
Supplier Number:  L105

Specifications

  • Description:
    Preprinted labels
  • Dimensions:
    50.8×101.6 mm (2×4")
  • Material:
    Semi-gloss paper
  • Thickness:
    5.85 mil.
  • Cat. No.:
    101376-632
  • Packaging:
    On roll
  • Legend:
    Protect From Freezing
  • Supplier no.:
    L105

Specifications

About this item

Protect From Freezing labels are great for calling out additional special handling instructions for temperature-sensitive packages.

Providing strong adhesion to corrugated boxes, this paper label alerts shipping and receiving personnel to special handling instructions.