Order Entry
United States
ContactUsLinkComponent
Combustible Liquid Labels, Labelmaster
Combustible Liquid Labels, Labelmaster
Catalog # 101377-718
Supplier:  Label Master
Combustible Liquid Labels, Labelmaster
Catalog # 101377-718
Supplier:  Label Master
Supplier Number:  HSLC4R

Specifications

  • Description:
    Combustible liquid labels
  • Dimensions:
    101.6×101.6 mm (4×4")
  • Material:
    Permanent vinyl
  • Thickness:
    11.2 mil.
  • Cat. No.:
    101377-718
  • Legend:
    Combustible Liquid
  • Supplier no.:
    HSLC4R

Specifications

About this item

Ensure shipping handlers' safety with Combustible Liquid labels.

  • Made of flexible, weather-resistant PVC-free vinyl
  • Features automotive-grade ink pigments
  • Specially formulated adhesives

These labels serve as a cautionary warning for materials that meet the definition of combustible in 49 CFR. They are discretionary for packaging less than 119 gallons.