Order Entry
United States
ContactUsLinkComponent
Shoulder Labels, Labelmaster
Shoulder Labels, Labelmaster
Catalog # 101378-070
Supplier:  Label Master
Shoulder Labels, Labelmaster
Catalog # 101378-070
Supplier:  Label Master
Supplier Number:  HDT38

Specifications

  • Description:
    Shoulder labels
  • Dimensions:
    50.8×133.4 mm (2×5.25")
  • Material:
    Permanent vinyl
  • Thickness:
    11.2 mil.
  • Cat. No.:
    101378-070
  • Legend:
    Carbon Dioxide UN1013
  • Supplier no.:
    HDT38

Specifications

About this item

These Shoulder labels are designed specifically for easy application to the tops of cylinders.

  • Automotive-grade ink pigments
  • Pre-printed with UN1013 Carbon Dioxide on exterior-grade, flexible vinyl
  • Withstands rough handling, extreme temperatures, UV exposure, moisture, scraping, and abrasion

These non-flammable gas UN1013 labels meet the DOT requirements in 49 CFR and the Compressed Gas Association's C-7 pamphlet.