Order Entry
United States
ContactUsLinkComponent
Inspected Labels, Labelmaster
Inspected Labels, Labelmaster
  101376-570
Inspected Labels, Labelmaster
  101376-570
 :  BT304

 

  • Description:
    Inspection labels
  • Dimensions:
    38.1×50.8 mm (1.5×2")
  • Material:
    Fluorescent orange paper
  • Thickness:
    7.1 mil.
  • Cat. No.:
    101376-570
  • Core size:
    3"
  • Legend:
    Inspected; Date__; By__
  • Supplier no.:
    BT304

 

 

Create a customized labeling solution for inspected materials with Inspected labels.

  • Made of paper
  • Strong adhesion to corrugated boxes
  • Bright fluorescent orange ink

Once filled in, these labels will instantly command attention wherever they're used and help with expedited shipping, production, and office work.