Attention This Container Hazardous When Empty Labels, Labelmaster
101376-253
- Description:Warning labels
- Dimensions:127×101.6 mm (5×4")
- Material:PVC-free film
- Thickness:7.1 mil.
- Cat. No.:101376-253
- Legend:Attention This Container Hazardous When Empty
- Supplier no.:L135A
Rest easy knowing that shipments will receive special handling with Attention This Container Hazardous When Empty labels.
- Constructed of rugged, flexible, and PVC-free vinyl
- Permanent adhesive for excellent adhesion to cardboard bo×es
These warehouse labels feature bold lettering for easy visibility. They can be used to e×pedite shipping, product, and office work.